Learn More
Biomatik Corporation PKR (Phospho-Thr446) anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41824
The PKR (Phospho-Thr446) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Synthetic peptide, Human: PKR around the phosphorylation site of Thr446. Applications: WB, ELISA. Gene ID: 5610. Gene Name: EIF2AK2. Alternative Names: Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2; eIF-2A protein kinase 2; Protein kinase RNA-activated; PKR; p68 kinase; P1/eIF-2A protein kinase. Swiss-Prot: P19525. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6020