Learn More
Biomatik Corporation SP1 (Phospho-Thr739) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41843
The SP1 (Phospho-Thr739) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: SP1 around the phosphorylation site of Thr739. Applications: WB, ELISA. Gene ID: 6667. Gene Name: SP1. Alternative Names: Transcription factor SP1; transcription factor Sp1; TSFP1. Swiss-Prot: P08047. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6022