Learn More
Biomatik Corporation Mmp7 anti-Rat, Mouse, Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07102
The Mmp7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat, Mouse, Human. Immunogen: Recombinant Rat Matrilysin protein (98-267AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Matrin Matrix metalloproteinase-7 Pump-1 protease Uterine metalloproteinase Mmp7 Mmp-7. UniProt: P50280. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6689