Learn More
Biomatik Corporation CASP1 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07116
The CASP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Caspase-1 protein (120-269AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. UniProt: P29466. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6703