Learn More
Biomatik Corporation TRAF6 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07123
The TRAF6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human TNF receptor-associated factor 6 protein (1-522AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:50-1:200, IF:1:50-1:500. Alternative Names: TNF receptor-associated factor 6, E3 ubiquitin-protein ligase TRAF6, Interleukin-1 signal transducer, RING finger protein 85, TRAF6, RNF85. UniProt: Q9Y4K3. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6710