Learn More
Biomatik Corporation LGALS1 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07134
The LGALS1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Galectin-1 protein (2-135AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, Galaptin, HBL, HPL, Lactose-binding lectin 1, Lectin galactoside, binding soluble 1, Putative MAPK-activating protein PM12, S-Lac lectin 1, LGALS1. UniProt: P09382. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6721