Learn More
Biomatik Corporation STAT6 (Phospho-Tyr641) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41854
The STAT6 (Phospho-Tyr641) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: STAT6 around the phosphorylation site of Tyr641. Applications: WB, IHC, ELISA. Gene ID: 6778. Gene Name: STAT6. Alternative Names: Signal transducer and activator of transcription 6; IL-4 Stat. Swiss-Prot: P42226. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6026