Learn More
Biomatik Corporation UBE2I anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07148
The UBE2I Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human SUMO-conjugating enzyme UBC9 protein (1-157AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: UBE2I, UBC9, UBCE9, SUMO-protein ligase, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, Ubiquitin-conjugating enzyme E2 I, Ubiquitin-protein ligase I, p18. UniProt: P63279. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6735