Learn More
Biomatik Corporation UCP3 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07152
The UCP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Mitochondrial uncoupling protein 3 protein (33-76AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Mitochondrial uncoupling protein 3, UCP 3, Solute carrier family 25 member 9, UCP3, SLC25A9. UniProt: P55916. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6739