Learn More
Biomatik Corporation PPP2CB anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07182
The PPP2CB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform protein (1-309AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: PP2A-beta. UniProt: P62714. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6769