Learn More
Biomatik Corporation GABARAPL2 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07188
The GABARAPL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Gamma-aminobutyric acid receptor-associated protein-like 2 protein (1-117AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:1000-1:2000. UniProt: P60520. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6775