Learn More
Biomatik Corporation PRDX4 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07195
The PRDX4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Peroxiredoxin-4 protein (38-271AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: Peroxiredoxin-4 (EC:1.11.1.15), PRDX4, Antioxidant enzyme AOE372, AOE37-2, Peroxiredoxin IV, Prx-IV, Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372. UniProt: Q13162. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6782