Learn More
Biomatik Corporation HIST1H3A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07196
The HIST1H3A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Histone H3.1 protein (108-136AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/, HIST1H3A, H3FA, AND, HIST1H3B, H3FL, AND, HIST1H3C, H3FC, AND, HIST1H3D, H3FB, AND, HIST1H3E, H3FD, AND, HIST1H3F, H3FI, AND, HIST1H3G, H3FH, AND, HIST1H3H, H3FK, AND, HIST1H3I, H3FF, AND, HIST1H3J, H3FJ. UniProt: P68431. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6783