Learn More
Biomatik Corporation YWHAH anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07214
The YWHAH Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human 14-3-3 protein eta protein (4-246AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Protein AS1, YWHAH, YWHA1. UniProt: Q04917. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6801