Learn More
Biomatik Corporation CCR6 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07230
The CCR6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human C-C chemokine receptor type 6 protein (22-374AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: C-C chemokine receptor type 6, C-C CKR-6, CC-CKR-6, CCR-6, Chemokine receptor-like 3, CKR-L3, DRY6, G-protein coupled receptor 29, GPR-CY4, GPRCY4, LARC receptor, CCR6, CKRL3, CMKBR6, GPR29, STRL22. UniProt: P51684. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6817