Learn More
Biomatik Corporation MAP3K7 (Phospho-Thr187) anti-Human, Murine, Rat, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA41945
The MAP3K7 (Phospho-Thr187) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine, Rat. Immunogen: Synthetic peptide, Human: MAP3K7 around the phosphorylation site of Thr187. Applications: WB, ELISA. Gene ID: 6885. Gene Name: MAP3K7. Alternative Names: M3K7; MAP3K7; Mitogen-activated protein kinase kinase kinase 7; TGF-beta- activated kinase 1; Transforming growth factor-beta-activated kinase 1; kinase TAK1. Swiss-Prot: O43318. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6035