Learn More
Biomatik Corporation PSMD7 anti-Human, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07240
The PSMD7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human 26S proteasome non-ATPase regulatory subunit 7 protein (1-324AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB:1:1000-1:5000, IHC:1:500-1:1000, IF:1:50-1:500, IP:1:200-1:2000. Alternative Names: 26S proteasome non-ATPase regulatory subunit 7, PSMD7, MOV34L, 26S proteasome regulatory subunit RPN8, 26S proteasome regulatory subunit S12, Mov34 protein homolog, Proteasome subunit p40. UniProt: P51665. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6827