Learn More
Biomatik Corporation blaNDM-1 anti-Klebsiella pneumoniae, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAC07248
The blaNDM-1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Klebsiella pneumoniae. Immunogen: Recombinant Klebsiella pneumoniae Beta-lactamase NDM-1 protein (29-270AA). Applications: ELISA. Alternative Names: NDM-1, Metallo-beta-lactamase NDM-1, blaNDM-1. UniProt: C7C422. Purification: >95%, Protein G purified. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6835