Learn More
Biomatik Corporation VE-Cadherin (Phospho-Tyr731) anti-Human, Murine, Rabbit Polyclonal Antibody, 100UG DFS Item
Manufacturer:  Biomatik Corporation CAA42011
The VE-Cadherin (Phospho-Tyr731) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Murine. Immunogen: Synthetic peptide, Human: VE-Cadherin around the phosphorylation site of Tyr731. Applications: WB, IHC, ELISA. Gene ID: 1003. Gene Name: CDH5. Alternative Names: 7B4 antigen; CADH5; Cadherin-5; CD144 antigen; Vascular endothelial-cadherin precursor; VE-cadherin. Swiss-Prot: P33151. Purification: Affinity Purified. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-6043