Learn More
Biomatik Corporation Recombinant Ferroportin (FPN), 100UG
Supplier:  Biomatik Corporation RPU41396
Encompass_Preferred
The purity of Recombinant Ferroportin (FPN) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Ferroportin. Alternative Names: SLC40A1; FPN1; HFE4; IREG1; MTP1; SLC11A3; Solute Carrier Family 40,Member 1; Iron-Regulated Transporter. UniProt: Q9NP59. Expression Region: Trp127-Val321. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 53.7 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5545