Learn More
Biomatik Corporation Recombinant Glial Fibrillary Acidic Protein (GFAP), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU41608
The purity of Recombinant Glial Fibrillary Acidic Protein (GFAP) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Glial Fibrillary Acidic Protein. Alternative Names: Intermediate Filament Protein. UniProt: P14136. Expression Region: Glu254-Glu374. Tag Info: two N-terminal Tags, His-tag and S-tag. Theoretical MW: 20.0 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5560