Learn More
Biomatik Corporation Recombinant Heat Shock 70kDa Protein 5 (HSPA5), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU41821
The purity of Recombinant Heat Shock 70kDa Protein 5 (HSPA5) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Heat Shock 70kDa Protein 5. Alternative Names: MIF2; BIP; GRP78; Immunoglobulin heavy chain-binding protein; Glucose Regulated Protein 78; Binding Immunoglobulin Protein; Endoplasmic reticulum lumenal Ca binding grp78. UniProt: P20029. Expression Region: Gln261-Glu470. Tag Info: N-terminal His-Tag. Theoretical MW: 24.7 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5573