Learn More
Biomatik Corporation Recombinant Hedgehog Homolog, Sonic (SHH), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU41844
The purity of Recombinant Hedgehog Homolog, Sonic (SHH) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Rattus norvegicus (Rat). Gene Name: Hedgehog Homolog, Sonic. Alternative Names: HHG1; HLP3; HPE3; SMMCI; SHHN; SHH-N. UniProt: Q63673. Expression Region: Gly28-Val192. Tag Info: N-terminal His-Tag. Theoretical MW: 20.4 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5574