Learn More
Biomatik Corporation Recombinant Inter Alpha-Globulin Inhibitor H4 (ITIH4), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU42095
The purity of Recombinant Inter Alpha-Globulin Inhibitor H4 (ITIH4) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Rattus norvegicus (Rat). Gene Name: Inter Alpha-Globulin Inhibitor H4. Alternative Names: IHRP; PK120; H4P; ITIHL1; Plasma Kallikrein-Sensitive Glycoprotein; Plasma kallikrein sensitive glycoprotein 120; Inter-alpha-trypsin inhibitor family heavy chain-related protein. UniProt: Q5EBC0. Expression Region: Thr478-Val722. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 58.7 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5598