Learn More
Biomatik Corporation Recombinant Interferon Gamma (IFNg), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU42143
The purity of Recombinant Interferon Gamma (IFNg) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Sus scrofa; Porcine (Pig). Gene Name: Interferon Gamma. Alternative Names: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. UniProt: P17803. Expression Region: Ser21-Lys166. Tag Info: N-terminal His-Tag. Theoretical MW: 18.7 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5609