Learn More
Biomatik Corporation Recombinant Interleukin 18 (IL18), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU42255
The purity of Recombinant Interleukin 18 (IL18) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Interleukin 18. Alternative Names: IGIF; IL-1g; IL1F4; IL1-F4; Interferon-Gamma-Inducing Factor; Interleukin-1 Family Member 4; Iboctadekin; Interleukin-1 Gamma. UniProt: Q14116. Expression Region: Tyr37-Asp193. Tag Info: N-terminal His-Tag. Theoretical MW: 19.5 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5615