Learn More
Biomatik Corporation Recombinant Monokine Induced By Interferon Gamma (MIg), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU42800
The purity of Recombinant Monokine Induced By Interferon Gamma (MIg) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Monokine Induced By Interferon Gamma. Alternative Names: CXCL9; CMK; Humig; MI-G; SCYB9; Crg-10; Chemokine (C-X-C motif) ligand 9; Small-inducible cytokine B9; Gamma-interferon-induced monokine. UniProt: P18340. Expression Region: Thr22-Thr126. Tag Info: two N-terminal Tags, His-tag and T7-tag. Theoretical MW: 16.0 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5664