Learn More
Biomatik Corporation Recombinant Pappalysin 2 (PAPPA2), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU43172
The purity of Recombinant Pappalysin 2 (PAPPA2) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Pappalysin 2. Alternative Names: PAPP-A2; PAPPE; PLAC3; Placenta-Specific 3; Pregnancy-associated plasma protein A2; Pregnancy-associated plasma protein E1. UniProt: Q9BXP8. Expression Region: Gly888-Ala1093. Tag Info: N-terminal His-Tag. Theoretical MW: 24.6 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5690