Learn More
Biomatik Corporation Recombinant Podocin (PDCN), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU43366
The purity of Recombinant Podocin (PDCN) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Rattus norvegicus (Rat). Gene Name: Podocin. Alternative Names: NPHS2; NPHS-2; Nephrosis 2; Idiopathic Steroid-Resistant. UniProt: Q8K4G9. Expression Region: Arg4-Ala100. Tag Info: N-terminal His-Tag. Theoretical MW: 11.8 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5703