Learn More
Biomatik Corporation Recombinant Proteinase 3 (PR3), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU43519
The purity of Recombinant Proteinase 3 (PR3) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Proteinase 3. Alternative Names: MBN; PRTN3; ACPA; AGP7; C-ANCA; MBT; P29; PRK; Myeloblastin; Serine Proteinase,Neutrophil,Wegener Granulomatosis Autoantigen; Protease K; Proteinase K; Endopeptidase K. UniProt: Q61096. Expression Region: Ala24-Leu249. Tag Info: N-terminal His-Tag. Theoretical MW: 26.3 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5711