Learn More
Biomatik Corporation Recombinant Selenoprotein P1, Plasma (SEPP1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU43740
The purity of Recombinant Selenoprotein P1, Plasma (SEPP1) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Selenoprotein P1, Plasma. Alternative Names: SEPP; SeP; SEP-P1; SELP. UniProt: P49908. Expression Region: Tyr60-Ser299. Tag Info: N-terminal His-Tag. Theoretical MW: 25.52 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5725