Learn More
Biomatik Corporation Recombinant Mouse H-2 class II histocompatibility antigen gamma chain (Cd74), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20020
The purity of Recombinant Mouse H-2 class II histocompatibility antigen gamma chain (Cd74), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Cd74. Alternative Names: Ia antigen-associated invariant chain; IiMHC class II-associated invariant chain; CD74. UniProt: P04441. Expression Region: 56-279aa. Sequence Info: Extracellular Domain. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 29.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4542