Learn More
Biomatik Corporation Recombinant Toll Like Receptor 8 (TLR8), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU44082
The purity of Recombinant Toll Like Receptor 8 (TLR8) is greater than 95%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Toll Like Receptor 8. Alternative Names: CD288. UniProt: Q9NR97. Expression Region: Leu844-Lys1039. Tag Info: two N-terminal Tags, His-tag and S-tag. Theoretical MW: 29.0 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5748