Learn More
Biomatik Corporation Recombinant Transferrin Receptor (TFR), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU44116
The purity of Recombinant Transferrin Receptor (TFR) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Transferrin Receptor. Alternative Names: CD71; TFR1; P90; TFRC; T9; Trfr; sTfR; Transferrin receptor protein 1, serum form. UniProt: Q62351. Expression Region: Arg572-Trp757. Tag Info: two N-terminal Tags, His-tag and S-tag. Theoretical MW: 27.4 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5750