Learn More
Biomatik Corporation Recombinant Troponin I Type 3, Cardiac (TNNI3), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU44188
The purity of Recombinant Troponin I Type 3, Cardiac (TNNI3) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Troponin I Type 3, Cardiac. Alternative Names: cTnI; TNNC1; CMH7; C-TnI; Troponin I Type 3; Cardiac troponin I. UniProt: P19429. Expression Region: Met1-Gly203. Tag Info: N-terminal His-Tag. Theoretical MW: 24.6 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5761