Learn More
Biomatik Corporation Recombinant Tryptase (TPS), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU44197
The purity of Recombinant Tryptase (TPS) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Tryptase. Alternative Names: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. UniProt: Q15661. Expression Region: Gly161-Tyr262. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 43.1 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5765