Learn More
Biomatik Corporation Recombinant Tumor Necrosis Factor Alpha (TNFa), 100UG
Supplier:  Biomatik Corporation RPU44218
Encompass_Preferred
The purity of Recombinant Tumor Necrosis Factor Alpha (TNFa) is greater than 95%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Tumor Necrosis Factor Alpha. Alternative Names: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. UniProt: P01375. Expression Region: Val77-Leu233. Tag Info: N-terminal His-Tag. Theoretical MW: 21.0 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5766