Learn More
Biomatik Corporation Recombinant Vascular Endothelial Growth Factor Receptor 1 (VEGFR1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU44332
The purity of Recombinant Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Sus scrofa; Porcine (Pig). Gene Name: Vascular Endothelial Growth Factor Receptor 1. Alternative Names: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Expression Region: Pro167-Ala424. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 58.89 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5779