Learn More
Biomatik Corporation Recombinant Toll Like Receptor 9 (TLR9), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU52016
The purity of Recombinant Toll Like Receptor 9 (TLR9) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Toll Like Receptor 9. Alternative Names: CD289. UniProt: Q9NR96. Expression Region: Thr28-Ser362. Tag Info: two N-terminal Tags, His-tag and T7-tag. Theoretical MW: 41.11 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5835