Learn More
Biomatik Corporation Recombinant Interleukin 3 (IL3), 100UG
Supplier:  Biomatik Corporation RPU52969
Encompass_Preferred
Recombinant Human Interleukin 3 (IL3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 3 (IL3). Target Synonyms: MCGF; MULTI-CSF; MCSF; M-CSF; Multiple; Multilineage-Colony-Stimulating Factor; Hematopoietic Growth Factor; P-Cell Stimulating Factor; Mast-Cell Growth Factor. Accession Number: P08700. Gene ID: 3562. Expression Region: Ala20~Phe152. Tag Info: N-terminal His and GST Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5860
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.