Learn More
Biomatik Corporation Eukaryotic Interleukin 4 (IL4), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU54798
The purity of Eukaryotic Interleukin 4 (IL4) is greater than 95%. Source: 293F cell. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Interleukin 4. Alternative Names: BSF1; BCGF1; B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. UniProt: P07750. Expression Region: His21-Ser140. Tag Info: N-terminal His Tag. Theoretical MW: 18/19 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5922