Learn More
Biomatik Corporation Recombinant Nerve Growth Factor (NGF), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU55176
The purity of Recombinant Nerve Growth Factor (NGF) is greater than 90%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Rattus norvegicus (Rat). Gene Name: Nerve Growth Factor. Alternative Names: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. UniProt: P25427. Expression Region: Glu19-Gly241 linked with LEHHHHHH. Tag Info: N-terminal His Tag. Theoretical MW: 33 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01%skl, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5933