Learn More
Biomatik Corporation Recombinant Mouse Acetylcholine receptor subunit alpha (Chrna1), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20024
The purity of Recombinant Mouse Acetylcholine receptor subunit alpha (Chrna1), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Chrna1. UniProt: P04756. Expression Region: 21-230aa. Sequence Info: Extracellular Domain. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 28.5kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4544