Learn More
Biomatik Corporation Recombinant Mouse Apolipoprotein E (Apoe), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20221
The purity of Recombinant Mouse Apolipoprotein E (Apoe) is greater than 90% as determined by SDS-PAGE. Source: Yeast. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Apoe. UniProt: P08226. Expression Region: 19-311aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 36kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4671