Learn More
Biomatik Corporation Recombinant Human Transcriptional enhancer factor TEF-1 (TEAD1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20246
The purity of Recombinant Human Transcriptional enhancer factor TEF-1 (TEAD1) is greater than 90% as determined by SDS-PAGE. Source: Yeast. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Homo sapiens (Human). Gene Name: TEAD1. Alternative Names: NTEF-1; Protein GT-IICTEA domain family member 1; TEAD-1Transcription factor 13; TCF-13. UniProt: P28347. Expression Region: 1-426aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-tagged. Theoretical MW: 49.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4679