Learn More
Biomatik Corporation Recombinant Mouse Placenta-expressed transcript 1 protein (Plet1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20280
The purity of Recombinant Mouse Placenta-expressed transcript 1 protein (Plet1) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Plet1. Alternative Names: Antigen mAgK114. UniProt: Q8VEN2. Expression Region: 28-218aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 36.1kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4697