Learn More
Biomatik Corporation Recombinant Helicobacter pylori Cytotoxicity-associated immunodominant antigen (cagA), partial, 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20295
The purity of Recombinant Helicobacter pylori Cytotoxicity-associated immunodominant antigen (cagA), partial is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Helicobacter pylori (Campylobacter pylori). Gene Name: cagA. Alternative Names: 120KDA protein. UniProt: P80200. Expression Region: 918-1147aa. Sequence Info: Partial. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 41.3kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4703