Learn More
Biomatik Corporation Recombinant Mouse Pancreatic triacylglycerol lipase (Pnlip), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20344
The purity of Recombinant Mouse Pancreatic triacylglycerol lipase (Pnlip) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Mus musculus (Mouse). Gene Name: Pnlip. Alternative Names:. PL. PTL. Pancreatic lipase. UniProt: Q6P8U6. Expression Region: 17-465aa. Sequence Info: Full Length of Mature Protein. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 65.8kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4736