Learn More
Biomatik Corporation Recombinant Bacillus subtilis Penicillin-binding protein 4 (pbpE), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20368
The purity of Recombinant Bacillus subtilis Penicillin-binding protein 4 (pbpE) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Bacillus subtilis (strain 168). Gene Name: pbpE. Alternative Names: PBP 4*. Alternative name (s): PBP 4A. Penicillin-binding protein E. UniProt: P32959. Expression Region: 1-451aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 67.4kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4752