Learn More
Biomatik Corporation Recombinant Helicobacter pylori DNA protection during starvation protein (dps), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPC20384
The purity of Recombinant Helicobacter pylori DNA protection during starvation protein (dps) is greater than 90% as determined by SDS-PAGE. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: Not tested. Species: Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori). Gene Name: dps. Alternative Names: Bacterioferritin. HP-NAP. Neutrophil-activating protein A. NAP A. UniProt: P43313. Expression Region: 1-144aa. Sequence Info: Full Length. Tag Info: N-terminal 6xHis-SUMO-tagged. Theoretical MW: 32.9kDa. Storage Buffer: Tris-based buffer, 50% glycerol. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-4765